This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308779_m1
See other PHTF1 GE Assays ›
Gene Symbol:
Phtf1
Gene Name:
putative homeodomain transcription factor 1
Gene Aliases:
AU041898, AW049785, Phft, Phtf
Chromosome Location:
Chr.3: 103968110 - 104007490 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308779_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Phtf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163467.1 NP_001156939.1 17 - 18 2293 142
NM_001163468.1 NP_001156940.1 16 - 17 2540 142
NM_001163469.1 NP_001156941.1 16 - 17 2564 142
NM_013629.2 NP_038657.2 17 - 18 2699 142
GenBank mRNA AJ133721.1 - 17 - 18 2293 142
AJ242864.1 - 17 - 18 2699 142
AK088714.1 - 16 - 17 2431 142
AK131629.1 - 2590 142
BC021147.1 - 17 - 18 2572 142
BC050825.1 - 16 - 17 2376 6518647 142
BC057340.1 - 16 - 17 3551 6827816 142
BC060177.1 - 16 - 17 3551 6827816 142
BC064826.1 - 2 - 3 207 142
CO041244.1 - 2 - 3 142

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001070.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Phtf1

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_001163467.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_001163467.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_001163468.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_001163468.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_001163469.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_001163469.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_013629.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 NM_013629.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008695.mm.1 TC0300001070.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_001163467.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_001163467.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_001163468.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_001163468.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_001163469.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_001163469.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_013629.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 NM_013629.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300008694.mm.1 TC0300001070.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline