This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308807_m1
See other LRIG2 GE Assays ›
Gene Symbol:
Lrig2
Gene Name:
leucine-rich repeats and immunoglobulin-like domains 2
Gene Aliases:
4632419I10Rik, BB096938
Chromosome Location:
Chr.3: 104453980 - 104511938 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308807_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lrig2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025067.1 NP_001020238.1 15 - 16 2692 119
GenBank mRNA BC094228.1 - 15 - 16 2629 30362610 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002648.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lrig2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

sensory perception of sound
regulation of platelet-derived growth factor receptor signaling pathway
innervation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300020645.mm.1 NM_001025067.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300020645.mm.1 TC0300002648.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300020646.mm.1 NM_001025067.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300020646.mm.1 TC0300002648.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0