This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309270_m1
See other 9430007A20RIK GE Assays ›
Gene Symbol:
9430007A20Rik
Gene Name:
RIKEN cDNA 9430007A20 gene
Gene Aliases:
Aadacl4, Gm1034
Chromosome Location:
Chr.4: 144519822 - 144529353 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309270_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9430007A20Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198662.3 NP_941064.2 3 - 4 510 67
GenBank mRNA AK034567.1 - 3 - 4 511 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001851.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene 9430007A20Rik

Panther Classification:

Molecular Function -

deacetylase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
molecular_function
phospholipase activity
prenylcysteine methylesterase activity
lipase activity
hydrolase activity
1-oxa-2-oxocycloheptane lactonase activity
sulfolactone hydrolase activity
3,4-dihydrocoumarin hydrolase activity
butyrolactone hydrolase activity
endosulfan lactone lactonase activity
L-ascorbate 6-phosphate lactonase activity
Ser-tRNA(Thr) hydrolase activity
Ala-tRNA(Pro) hydrolase activity
Cys-tRNA(Pro) hydrolase activity
Ser(Gly)-tRNA(Ala) hydrolase activity
all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
retinyl-palmitate esterase activity
mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
methyl indole-3-acetate esterase activity
methyl salicylate esterase activity
methyl jasmonate esterase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156953 NM_198662.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2156953 AK034567.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193495 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751538 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820224 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104935 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839054 AF408435.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839054 BC005475.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839054 NM_145150.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24797 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107935 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156953 NM_198662.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2156953 AK034567.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378858 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378857 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378866 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378840 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378839 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378831 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378867 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378844 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378860 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378864 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378859 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378863 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378843 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378870 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378838 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378871 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378850 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378836 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378869 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378854 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378852 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378832 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378868 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378835 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23378856 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107935 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05437353 - -
Whole Mouse Genome G4122A A_51_P489092 BC148288.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P489092 NM_198662.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P489092 AK034567.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139186 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107935 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03392603 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03586815 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03586824 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03586818 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03586819 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378868 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378854 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378902 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378880 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378874 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378834 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378840 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378918 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378890 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378856 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378912 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378858 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378909 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378838 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378861 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378838 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378854 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03549137 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378880 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378836 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378858 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378852 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378876 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378886 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03586818 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03414958 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378835 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03250515 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378887 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04075904 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378868 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378840 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378863 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378860 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378890 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378879 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378864 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03414954 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03647194 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378859 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03586820 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378850 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378888 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378874 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378843 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378831 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03334956 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378847 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378872 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378869 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378866 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378902 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378839 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378841 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378870 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03626316 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03355776 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23378834 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10510122 - -
Affymetrix Mouse Gene 1.0 ST Array 10510124 - -
Affymetrix Mouse Gene 1.0 ST Array 10510121 - -
Affymetrix Mouse Gene 1.0 ST Array 10510123 - -
Affymetrix Mouse Gene 1.1 ST Array 10510120 - -
Affymetrix Mouse Clariom S Array PSR0400015202.mm.1 NM_198662.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400015202.mm.1 NM_198662.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400015202.mm.1 TC0400001851.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 878641 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4590008 - -
Illumina mouse-6 v1.0 expression beadchip 4590008 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004590008 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2723726 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2723726 - -
mouse-6 v1.1 (Illumina) scl0381572.4_223-S - -
Illumina Sentrix mouse-6.v1 bead array 4590008 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0381572.4_223-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590008 - -
MouseRef-8 v2 Expression BeadChip ILMN_2723726 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2723726 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0381572.4_223-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline