This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309303_m1
See other SLC8A3 GE Assays ›
Gene Symbol:
Slc8a3
Gene Name:
solute carrier family 8 (sodium/calcium exchanger), member 3
Gene Aliases:
AW742262, Ncx3
Chromosome Location:
Chr.12: 81197915 - 81333728 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309303_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc8a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080440.3 NP_536688.2 4 - 5 2511 73
GenBank mRNA AF453257.1 - 4 - 5 2512 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200002012.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc8a3

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
sodium ion transport
metal ion transport
sodium ion transmembrane transport
calcium ion transport into cytosol
calcium ion transmembrane transport
cellular response to cAMP
calcium ion export from cell
calcium ion import into cell
calcium:sodium antiporter activity
calcium:cation antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200013845.mm.1 NM_001167920.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013845.mm.1 NM_001167920.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013845.mm.1 NM_080440.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200013845.mm.1 NM_080440.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200013845.mm.1 TC1200002012.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0