This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309662_m1
See other UNC45B GE Assays ›
Gene Symbol:
Unc45b
Gene Name:
unc-45 homolog B (C. elegans)
Gene Aliases:
AA445617, Cmya4, D230041A13Rik, Unc45
Chromosome Location:
Chr.11: 82911253 - 82943406 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309662_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc45b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178680.4 NP_848795.3 14 - 15 2066 68
GenBank mRNA AF539793.1 - 13 - 14 1965 68
AK052294.1 - 14 - 15 1993 68
AK081821.1 - 14 - 15 2005 68
AK084418.1 - 14 - 15 2034 68
AK168968.1 - 14 - 15 2035 68
BC084585.1 - 14 - 15 1999 30429260 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001235.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc45b

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
muscle organ development
cell differentiation
chaperone-mediated protein folding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100011593.mm.1 NM_178680.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100011593.mm.1 NM_178680.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100011593.mm.1 TC1100001235.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100011592.mm.1 NM_178680.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100011592.mm.1 NM_178680.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100011592.mm.1 TC1100001235.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0