This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309666_m1
See other UNC45B GE Assays ›
Gene Symbol:
Unc45b
Gene Name:
unc-45 homolog B (C. elegans)
Gene Aliases:
AA445617, Cmya4, D230041A13Rik, Unc45
Chromosome Location:
Chr.11: 82911253 - 82943406 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309666_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc45b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178680.4 NP_848795.3 18 - 19 2481 86
GenBank mRNA AF539793.1 - 17 - 18 2380 86
AK052294.1 - 18 - 19 2408 86
AK081821.1 - 18 - 19 2420 86
AK084418.1 - 18 - 19 2449 86
AK168968.1 - 18 - 19 2450 86
BC084585.1 - 18 - 19 2414 30429260 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001235.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc45b

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
muscle organ development
cell differentiation
chaperone-mediated protein folding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100011597.mm.1 NM_178680.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100011597.mm.1 NM_178680.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100011597.mm.1 TC1100001235.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0