This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01309878_m1
See other KSR1 GE Assays ›
Gene Symbol:
Ksr1
Gene Name:
kinase suppressor of ras 1
Gene Aliases:
AW492498, B-KSR1, D11Bhm183e, D11Bhm184e, Ksr, RSU2, hb
Chromosome Location:
Chr.11: 79013440 - 79146554 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01309878_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ksr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013571.2 NP_038599.1 9 - 10 1333 97
GenBank mRNA AK036566.1 - 9 - 10 1389 97
AK137532.1 - 8 - 9 1070 97
AK164833.1 - 9 - 10 1455 97
U43585.1 - 9 - 10 1340 97
X81634.1 - 9 - 10 1115 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003315.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ksr1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
intracellular signal transduction
positive regulation of MAPK cascade
protein kinase activity
MAP-kinase scaffold activity
protein binding
ATP binding
protein C-terminus binding
transferase activity, transferring phosphorus-containing groups
mitogen-activated protein kinase kinase binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100030523.mm.1 NM_013571.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100030523.mm.1 TC1100003315.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline