This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310027_m1
See other RPS6KB1 GE Assays ›
Gene Symbol:
Rps6kb1
Gene Name:
ribosomal protein S6 kinase, polypeptide 1
Gene Aliases:
2610318I15Rik, 4732464A07Rik, 70kDa, AA959758, AI256796, AI314060, S6K1, p70/85s6k, p70s6k
Chromosome Location:
Chr.11: 86498869 - 86544807 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310027_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps6kb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114334.1 NP_001107806.1 10 - 11 1080 66
GenBank mRNA AK012045.1 - 5 - 6 459 66
AK032724.1 - 10 - 11 1042 66
AK135716.1 - 10 - 11 1048 66
AK163258.1 - 10 - 11 1050 66
AK214758.1 - 2 - 3 66
BC038491.1 - 10 - 11 1053 4010710 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003447.mm.1

Set Membership:

Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
behavioral fear response
regulation of translation
protein phosphorylation
apoptotic process
cell cycle
signal transduction
germ cell development
positive regulation of smooth muscle cell migration
phosphorylation
TOR signaling
response to insulin
cellular response to insulin stimulus
response to tumor necrosis factor
negative regulation of apoptotic process
protein kinase B signaling
positive regulation of translation
positive regulation of mitotic cell cycle
positive regulation of translational initiation
regulation of glucose import
negative regulation of insulin receptor signaling pathway
positive regulation of skeletal muscle tissue growth
positive regulation of smooth muscle cell proliferation
cellular response to growth factor stimulus
cellular response to organic cyclic compound
negative regulation of extrinsic apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ribosomal protein S6 kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
PDZ domain binding
peptide binding
protein phosphatase 2A binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100031652.mm.1 NM_001114334.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100031652.mm.1 NM_001114334.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100031652.mm.1 TC1100003447.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline