This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310130_g1
See other RANGAP1 GE Assays ›
Gene Symbol:
Rangap1
Gene Name:
RAN GTPase activating protein 1
Gene Aliases:
C79654, Fug1, mKIAA1835
Chromosome Location:
Chr.15: 81704248 - 81729919 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310130_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rangap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK088768.1 - 11 - 12 1429 93
AK129452.1 - 11 - 12 1436 93
AK186071.1 - 1 - 2 39 93
BC052862.1 - 11 - 12 1430 30063509 93

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500015132.mm.1 NM_001146174.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015132.mm.1 NM_001146174.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015132.mm.1 NM_011241.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015132.mm.1 NM_011241.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500015132.mm.1 TC1500001927.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0