This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310132_m1
See other RANGAP1 GE Assays ›
Gene Symbol:
Rangap1
Gene Name:
RAN GTPase activating protein 1
Gene Aliases:
C79654, Fug1, mKIAA1835
Chromosome Location:
Chr.15: 81704248 - 81729919 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310132_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rangap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146174.1 NP_001139646.1 13 - 14 1735 73
NM_011241.4 NP_035371.4 14 - 15 1783 73
GenBank mRNA AK042930.1 - 13 - 14 1701 73
AK088768.1 - 13 - 14 1657 73
AK129452.1 - 13 - 14 1664 73
AK133905.1 - 13 - 14 1703 73
AK145459.1 - 12 - 13 1338 73
AK146670.1 - 14 - 15 1743 73
AK159009.1 - 13 - 14 1688 73
AK159525.1 - 13 - 14 1700 73
AK159930.1 - 13 - 14 1691 73
BC014855.1 - 13 - 14 1691 3157225 73
BC052862.1 - 13 - 14 1658 30063509 73
BC066213.1 - 13 - 14 1699 30466595 73
BC071200.1 - 14 - 15 1899 5032610 73
U08110.1 - 14 - 15 1766 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001927.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rangap1

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
negative regulation of protein export from nucleus
cellular response to peptide hormone stimulus
GTPase activator activity
Ran GTPase binding
ubiquitin protein ligase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500015131.mm.1 NM_001146174.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015131.mm.1 NM_001146174.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015131.mm.1 NM_011241.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015131.mm.1 NM_011241.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500015131.mm.1 TC1500001927.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500015130.mm.1 NM_001146174.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015130.mm.1 NM_001146174.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015130.mm.1 NM_011241.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015130.mm.1 NM_011241.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500015130.mm.1 TC1500001927.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0