This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310472_m1
See other NEIL2 GE Assays ›
Gene Symbol:
Neil2
Gene Name:
nei like 2 (E. coli)
Gene Aliases:
Gm1212, NEH2
Chromosome Location:
Chr.14: 63182445 - 63194244 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310472_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Neil2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201610.2 NP_963904.2 4 - 5 792 87
GenBank mRNA AY518221.1 - 4 - 5 808 87
BC107389.2 - 2 - 3 87
BC115709.1 - 4 - 5 792 87
BC115762.1 - 4 - 5 792 40058279 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002216.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Neil2

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
metabolic process
purine-specific mismatch base pair DNA N-glycosylase activity
nucleic acid binding
DNA binding
damaged DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
microtubule binding
zinc ion binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
hydrolase activity, hydrolyzing N-glycosyl compounds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
metal ion binding
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099418 AY518221.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2099418 NM_201610.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362171 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927407 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321414 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421565 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435736 AK046969.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435736 BC080293.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435736 BC108419.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435736 NM_001033286.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167152 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666067 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2763 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135298 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099418 AY518221.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2099418 NM_201610.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659727 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659754 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659743 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659664 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659721 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659720 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659712 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659766 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659662 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659730 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659747 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659719 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659736 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659757 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659725 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659711 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659716 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659735 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659724 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659710 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659718 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659666 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659745 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659762 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659769 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659749 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659760 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659714 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659708 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659709 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659746 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659738 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659733 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659756 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659723 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659742 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659731 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659728 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659759 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659668 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659768 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659740 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659737 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659663 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659751 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29659763 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02866362 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135298 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02866376 - -
Whole Mouse Genome G4122A A_52_P498396 AY518221.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P498396 NM_201610.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106300 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135298 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11739185 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11739198 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11739179 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11739194 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11739184 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11739203 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11739209 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659669 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659712 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659737 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659745 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659746 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659743 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659751 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659689 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659735 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659740 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659718 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659666 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659698 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659725 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29659720 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11680438 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659727 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11660272 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659721 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659724 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11747670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11660277 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659708 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659760 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11428426 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659709 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659730 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659738 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659766 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11731129 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659720 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659768 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659716 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659756 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11731128 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659706 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659762 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659672 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659757 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11788385 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659755 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659701 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659669 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659665 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659710 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659751 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11739206 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11731121 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659746 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659723 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659694 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11578655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659740 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11578656 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11660268 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659735 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659679 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659745 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659731 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659725 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11520781 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659742 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659684 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11428412 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659664 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11788387 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659677 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659754 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659714 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11788380 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659719 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659733 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659686 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659743 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11578646 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11792997 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11739194 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11278889 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659705 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11660269 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11428411 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29659736 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10420738 - -
Affymetrix Mouse Gene 1.0 ST Array 10420739 - -
Affymetrix Mouse Gene 1.0 ST Array 10420743 - -
Affymetrix Mouse Gene 1.0 ST Array 10420745 - -
Affymetrix Mouse Gene 1.0 ST Array 10420744 - -
Affymetrix Mouse Gene 1.0 ST Array 10420742 - -
Affymetrix Mouse Gene 1.0 ST Array 10420746 - -
Affymetrix Mouse Gene 1.0 ST Array 10420740 - -
Affymetrix Mouse Gene 1.0 ST Array 10420737 - -
Affymetrix Mouse Gene 1.0 ST Array 10420741 - -
Affymetrix Mouse Gene 1.1 ST Array 10420736 - -
Affymetrix Mouse Clariom S Array PSR1400017156.mm.1 NM_201610.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017156.mm.1 NM_201610.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400017156.mm.1 TC1400002216.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017153.mm.1 NM_201610.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017153.mm.1 NM_201610.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400017153.mm.1 TC1400002216.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017155.mm.1 NM_201610.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017155.mm.1 NM_201610.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400017155.mm.1 TC1400002216.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017154.mm.1 NM_201610.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017154.mm.1 NM_201610.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400017154.mm.1 TC1400002216.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017151.mm.1 NM_201610.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017151.mm.1 NM_201610.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400017151.mm.1 TC1400002216.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017152.mm.1 NM_201610.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017152.mm.1 NM_201610.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400017152.mm.1 TC1400002216.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 328153 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 3780010 - -
Illumina mouse-6 v1.0 expression beadchip 3780010 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003780010 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613131 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2613131 - -
mouse-6 v1.1 (Illumina) scl0382913.1_100-S - -
Illumina Sentrix mouse-6.v1 bead array 3780010 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0382913.1_100-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780010 - -
MouseRef-8 v2 Expression BeadChip ILMN_2613131 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613131 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0382913.1_100-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0