This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01310681_m1
See other GTF2F2 GE Assays ›
Gene Symbol:
Gtf2f2
Gene Name:
general transcription factor IIF, polypeptide 2
Gene Aliases:
1110031C13Rik, AI326315
Chromosome Location:
Chr.14: 75896937 - 76010865 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01310681_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2f2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026816.3 NP_081092.1 5 - 6 503 76
GenBank mRNA AK144858.1 - 5 - 6 504 76
BC027173.1 - 5 - 6 499 3986466 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002433.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2f2

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
positive regulation of transcription elongation from RNA polymerase II promoter
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
positive regulation of transcription from RNA polymerase II promoter
nucleotide binding
DNA binding
catalytic activity
helicase activity
protein binding
ATP binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400018847.mm.1 NM_026816.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018847.mm.1 NM_026816.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400018847.mm.1 TC1400002433.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400018849.mm.1 NM_026816.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018849.mm.1 NM_026816.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400018849.mm.1 TC1400002433.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0