This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01311282_m1
See other SMARCA1 GE Assays ›
Gene Symbol:
Smarca1
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1
Gene Aliases:
5730494M04Rik, Snf2l
Chromosome Location:
Chr.X: 47809370 - 47892931 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01311282_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarca1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290708.1 NP_001277637.1 15 - 16 2232 83
NM_053123.4 NP_444353.3 15 - 16 2232 83
GenBank mRNA AF325920.1 - 15 - 16 2239 83
AK030741.1 - 15 - 16 2199 83
AK032978.1 - 15 - 16 2201 83
BC016071.1 - 15 - 16 2209 83
BC057115.1 - 15 - 16 2232 6406330 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002154.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarca1

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

DNA strand renaturation
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
chromatin modification
neuron differentiation
ATP-dependent chromatin remodeling
positive regulation of transcription, DNA-templated
regulation of neural precursor cell proliferation
nucleotide binding
nucleic acid binding
DNA binding
chromatin binding
helicase activity
ATP binding
DNA-dependent ATPase activity
hydrolase activity
hydrolase activity, acting on acid anhydrides
hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
ATPase activity
nucleosome binding
annealing helicase activity
nucleosome-dependent ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00013601.mm.1 NM_053123.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00013601.mm.1 NM_053123.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00013601.mm.1 TC0X00002154.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0