This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01311377_g1
See other ARHGAP36 GE Assays ›
Gene Symbol:
Arhgap36
Gene Name:
Rho GTPase activating protein 36
Gene Aliases:
1100001E04Rik, rho-gap
Chromosome Location:
Chr.X: 49448328 - 49500250 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01311377_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap36
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081123.1 NP_001074592.1 3 - 4 444 78
GenBank mRNA AB073967.1 - 3 - 4 311 78
AK048167.1 - 3 - 4 332 78
BC090672.1 - 3 - 4 415 30941825 78
BC145645.1 - 3 - 4 410 40130948 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000443.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00002975.mm.1 NM_001081123.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00002975.mm.1 NM_001081123.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00002975.mm.1 TC0X00000443.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0