Assay Details
| Gene Symbol: Sccpdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178653.3 | NP_848768.1 | 8 - 9 | 1057 | 129 | |
| GenBank mRNA | AK042919.1 | - | 8 - 9 | 945 | 129 | |
| AK046033.1 | - | 8 - 9 | 1058 | 129 | ||
| AK046468.1 | - | 8 - 9 | 1045 | 129 | ||
| AK050554.1 | - | 8 - 9 | 1006 | 129 | ||
| AK077695.1 | - | 8 - 9 | 1007 | 129 | ||
| AK082803.1 | - | 8 - 9 | 942 | 129 | ||
| AK086448.1 | - | 8 - 9 | 1080 | 129 | ||
| AK086632.1 | - | 8 - 9 | 130 | |||
| AK166018.1 | - | 8 - 9 | 969 | 129 | ||
| BC025803.1 | - | 8 - 9 | 1054 | 5100552 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sccpdh
Genomic Map