This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01311547_m1
See other RPS6KC1 GE Assays ›
Gene Symbol:
Rps6kc1
Gene Name:
ribosomal protein S6 kinase polypeptide 1
Gene Aliases:
AA682037, B130003F20Rik, C80612, Rpk118
Chromosome Location:
Chr.1: 190772879 - 190912139 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01311547_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps6kc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178775.4 NP_848890.3 1 - 2 219 105
GenBank mRNA AK030313.1 - 197 105
AK149858.1 - 1 - 2 184 105
BB619353.1 - 1 - 2 220 105
CJ045347.1 - 1 - 2 220 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003795.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps6kc1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
signal transduction
phosphorylation
nucleotide binding
molecular_function
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100031027.mm.1 NM_178775.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100031027.mm.1 NM_178775.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100031027.mm.1 TC0100003795.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100031028.mm.1 NM_178775.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100031028.mm.1 NM_178775.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100031028.mm.1 TC0100003795.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline