Assay Details
| Gene Symbol: Ttll7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014905.1 | - | 8 - 9 | 1250 | 93 | |
| AM690750.1 | - | 7 - 8 | 891 | 93 | ||
| AM690751.1 | - | 7 - 8 | 891 | 93 | ||
More Information
Set Membership: |
|

Ttll7
Genomic Map