This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312422_m1
See other MCEE GE Assays ›
Gene Symbol:
Mcee
Gene Name:
methylmalonyl CoA epimerase
Gene Aliases:
1110007A04Rik
Chromosome Location:
Chr.7: 64392612 - 64412121 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312422_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mcee
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028626.1 NP_082902.1 1 - 2 74 91
GenBank mRNA AK003511.1 - 1 - 2 71 91
BC038157.1 - 1 - 2 74 1479934 91

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK003511.1, BC038157.1, NM_028626.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mcee

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

L-methylmalonyl-CoA metabolic process
methylmalonyl-CoA epimerase activity
isomerase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700008953.mm.1 TC0700000979.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700008953.mm.1 NM_028626.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700008954.mm.1 TC0700000979.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700008954.mm.1 NM_028626.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700008955.mm.1 TC0700000979.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700008955.mm.1 NM_028626.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0