This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312427_m1
See other APBA2 GE Assays ›
Gene Symbol:
Apba2
Gene Name:
amyloid beta (A4) precursor protein-binding, family A, member 2
Gene Aliases:
X11-like, X11L, XllL, mXllL
Chromosome Location:
Chr.7: 64501706 - 64753878 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312427_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apba2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291166.1 NP_001278095.1 5 - 6 1365 62
NM_001291167.1 NP_001278096.1 4 - 5 1311 62
NM_007461.2 NP_031487.1 5 - 6 1362 62
GenBank mRNA BC057620.1 - 5 - 6 1362 6402377 62
BC060269.1 - 2 - 3 1065 6810117 62
L34676.1 - 2 - 3 824 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000980.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apba2

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
transport
chemical synaptic transmission
locomotory behavior
regulation of gene expression
protein transport
multicellular organism growth
beta-amyloid binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700008968.mm.1 TC0700000980.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700008968.mm.1 NM_007461.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008968.mm.1 NM_007461.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700008967.mm.1 TC0700000980.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700008967.mm.1 NM_007461.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700008967.mm.1 NM_007461.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0