This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01312589_m1
See other RPF1 GE Assays ›
Gene Symbol:
Rpf1
Gene Name:
ribosome production factor 1 homolog (S. cerevisiae)
Gene Aliases:
2210420E24Rik, 2310066N05Rik, Bxdc5
Chromosome Location:
Chr.3: 146506344 - 146521431 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01312589_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027332.3 NP_081608.1 2 - 3 315 102
NM_027371.3 NP_081647.2 2 - 3 315 102
GenBank mRNA AK008988.1 - 2 - 3 304 102
AK010079.1 - 2 - 3 312 102
AK162073.1 - 2 - 3 318 102
AK171035.1 - 2 - 3 296 102
AK171079.1 - 2 - 3 304 102
BC056165.1 - 2 - 3 266 5694049 102
BC089369.1 - 2 - 3 300 30550759 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003101.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpf1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
maturation of 5.8S rRNA
maturation of LSU-rRNA
rRNA processing
biological_process
ribosome biogenesis
RNA binding
rRNA binding
rRNA primary transcript binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300024400.mm.1 NM_027332.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024400.mm.1 NM_027332.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300024400.mm.1 NM_027371.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024400.mm.1 NM_027371.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300024400.mm.1 TC0300003101.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0