This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01313036_m1
See other PRSS55 GE Assays ›
Gene Symbol:
Prss55
Gene Name:
protease, serine 55
Gene Aliases:
4933401F05Rik
Chromosome Location:
Chr.14: 64075435 - 64085433 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01313036_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081063.1 NP_001074532.1 2 - 3 279 66
GenBank mRNA AK016601.1 - 1 - 2 313 66
BC115562.2 - 2 - 3 280 40101914 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002228.mm.1

Set Membership:

Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
biological_process
molecular_function
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017255.mm.1 NM_001081063.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017255.mm.1 NM_001081063.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017255.mm.1 TC1400002228.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017258.mm.1 NM_001081063.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017258.mm.1 NM_001081063.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017258.mm.1 TC1400002228.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017256.mm.1 NM_001081063.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017256.mm.1 NM_001081063.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017256.mm.1 TC1400002228.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017257.mm.1 NM_001081063.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017257.mm.1 NM_001081063.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017257.mm.1 TC1400002228.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline