This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01313233_m1
See other SEC24B GE Assays ›
Gene Symbol:
Sec24b
Gene Name:
Sec24 related gene family, member B (S. cerevisiae)
Gene Aliases:
AI605202, SEC24
Chromosome Location:
Chr.3: 129982764 - 130061573 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01313233_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec24b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207209.2 NP_997092.1 4 - 5 1364 57
GenBank mRNA AK150076.1 - 3 - 4 441 57
BC046556.1 - 4 - 5 1196 57
BC046776.1 - 4 - 5 1364 5706007 57
BC057355.1 - 4 - 5 1225 6833698 57

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec24b

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
auditory receptor cell morphogenesis
outflow tract morphogenesis
cochlear nucleus development
aorta morphogenesis
auditory receptor cell stereocilium organization
lung morphogenesis
lung lobe morphogenesis
coronary artery morphogenesis
pulmonary artery morphogenesis
cardiovascular system development
regulation of establishment of planar polarity involved in neural tube closure
regulation of cargo loading into COPII-coated vesicle
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0