This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01313648_m1
See other ELP3 GE Assays ›
Gene Symbol:
Elp3
Gene Name:
elongator acetyltransferase complex subunit 3
Gene Aliases:
2610507P14Rik, KAT9
Chromosome Location:
Chr.14: 65530446 - 65593112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01313648_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253812.1 NP_001240741.1 7 - 8 784 115
NM_028811.3 NP_083087.1 7 - 8 727 115
NR_045599.1 7 - 8 727 115
GenBank mRNA AK012072.1 - 7 - 8 665 115
AK088457.1 - 7 - 8 748 115
AK150058.1 - 7 - 8 665 115
BC026461.1 - 5 - 6 379 115
BC037470.1 - 1 - 2 81 115
BC048822.1 - 2 - 3 232 115
BC057453.1 - 7 - 8 667 3969625 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002259.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Elp3

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
tRNA wobble uridine modification
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
nervous system development
central nervous system development
histone acetylation
positive regulation of cell migration
histone H3 acetylation
histone H4 acetylation
regulation of protein kinase activity
RNA polymerase II core binding
catalytic activity
histone acetyltransferase activity
N-acetyltransferase activity
phosphorylase kinase regulator activity
H3 histone acetyltransferase activity
H4 histone acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
metal ion binding
iron-sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017407.mm.1 NM_001253812.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017407.mm.1 NM_001253812.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017407.mm.1 NM_028811.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017407.mm.1 NM_028811.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017407.mm.1 NR_045599.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017407.mm.1 NR_045599.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017407.mm.1 TC1400002259.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017408.mm.1 NM_001253812.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017408.mm.1 NM_001253812.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017408.mm.1 NM_028811.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017408.mm.1 NM_028811.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017408.mm.1 NR_045599.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017408.mm.1 NR_045599.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017408.mm.1 TC1400002259.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400017406.mm.1 NM_001253812.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017406.mm.1 NM_001253812.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017406.mm.1 NM_028811.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017406.mm.1 NM_028811.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017406.mm.1 NR_045599.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017406.mm.1 NR_045599.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017406.mm.1 TC1400002259.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0