This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01313812_m1
See other EPHX2 GE Assays ›
Gene Symbol:
Ephx2
Gene Name:
epoxide hydrolase 2, cytoplasmic
Gene Aliases:
CEH, Eph2, SEH, sEP
Chromosome Location:
Chr.14: 66084372 - 66124522 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01313812_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ephx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271402.1 NP_001258331.1 2 - 3 354 69
NM_007940.4 NP_031966.2 2 - 3 274 69
GenBank mRNA AK002415.1 - 2 - 3 324 69
AK142714.1 - 2 - 3 247 69
BC015087.1 - 2 - 3 250 4218917 69
L05781.1 - 2 - 3 187 69
Z37107.1 - 2 - 3 204 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001271402.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ephx2

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin production involved in inflammatory response
lipid metabolic process
inflammatory response
metabolic process
response to toxic substance
positive regulation of gene expression
dephosphorylation
sensory perception of pain
aromatic compound catabolic process
cholesterol homeostasis
linoleic acid metabolic process
positive regulation of blood pressure
stilbene catabolic process
phospholipid dephosphorylation
regulation of cholesterol metabolic process
magnesium ion binding
catalytic activity
epoxide hydrolase activity
receptor binding
toxic substance binding
hydrolase activity
phosphatase activity
10-hydroxy-9-(phosphonooxy)octadecanoate phosphatase activity
lipid phosphatase activity
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017503.mm.1 NM_001271402.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017503.mm.1 NM_001271403.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017503.mm.1 NM_001271421.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017503.mm.1 NM_007940.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400017503.mm.1 TC1400002266.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline