This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01313907_g1
See other SDR39U1 GE Assays ›
Gene Symbol:
Sdr39u1
Gene Name:
short chain dehydrogenase/reductase family 39U, member 1
Gene Aliases:
2310014G06Rik
Chromosome Location:
Chr.14: 55897285 - 55900281 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01313907_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdr39u1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082975.1 NP_001076444.1 1 - 2 41 63
GenBank mRNA AK049709.1 - 1 - 2 35 63
BY127480.1 - 1 - 2 41 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK049709.1, NM_001082975.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
oxidation-reduction process
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
molecular_function
catalytic activity
aldo-keto reductase (NADP) activity
isocitrate dehydrogenase activity
mevaldate reductase activity
gluconate dehydrogenase activity
steroid dehydrogenase activity
oxidoreductase activity
epoxide dehydrogenase activity
5-exo-hydroxycamphor dehydrogenase activity
2-hydroxytetrahydrofuran dehydrogenase activity
acetoin dehydrogenase activity
phenylcoumaran benzylic ether reductase activity
D-xylose:NADP reductase activity
L-arabinose:NADP reductase activity
D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
steroid dehydrogenase activity, acting on the CH-CH group of donors
(R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
3-hydroxymenthone dehydrogenase activity
very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
dihydrotestosterone 17-beta-dehydrogenase activity
(R)-2-hydroxyisocaproate dehydrogenase activity
L-arabinose 1-dehydrogenase (NADP+) activity
L-xylulose reductase (NAD+) activity
3-ketoglucose-reductase activity
coenzyme binding
(R)-2-hydroxyglutarate dehydrogenase activity
D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline