This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01313992_g1
See other HSD11B1 GE Assays ›
Gene Symbol:
Hsd11b1
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 193221640 - 193264049 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01313992_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd11b1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044751.1 NP_001038216.1 3 - 4 427 103
NM_008288.2 NP_032314.2 4 - 5 453 103
GenBank mRNA AI099306.1 - 3 - 4 430 103
AK149518.1 - 3 - 4 425 103
AK165212.1 - 3 - 4 425 103
AK167419.1 - 2 - 3 377 103
BC132364.1 - 4 - 5 393 40130641 103
BC138780.1 - 4 - 5 393 8861802 103
BC145415.1 - 4 - 5 393 9053973 103
DQ089001.1 - 3 - 4 329 103
S75207.1 - 3 - 4 374 103
X83202.1 - 4 - 5 456 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003836.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsd11b1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
glucocorticoid biosynthetic process
glucocorticoid catabolic process
metabolic process
steroid metabolic process
lung development
regulation of pentose-phosphate shunt
oxidation-reduction process
11-beta-hydroxysteroid dehydrogenase [NAD(P)] activity
steroid binding
oxidoreductase activity
NADP binding
11-beta-hydroxysteroid dehydrogenase (NADP+) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100031413.mm.1 NM_001044751.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100031413.mm.1 NM_001044751.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100031413.mm.1 NM_008288.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100031413.mm.1 NM_008288.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100031413.mm.1 TC0100003836.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0