Assay Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153137.4 | NP_694777.3 | 6 - 7 | 991 | 72 | |
| GenBank mRNA | AK031400.1 | - | 6 - 7 | 931 | 72 | |
| AK041994.1 | - | 6 - 7 | 992 | 72 | ||
More Information
Set Membership: |
|

Traf3ip3
Genomic Map