| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700031401.mm.1 |
NR_046006.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700031401.mm.1 |
NR_046006.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700031401.mm.1 |
NR_046007.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700031401.mm.1 |
NR_046007.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700031401.mm.1 |
TC0700003722.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700031405.mm.1 |
NR_046007.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700031405.mm.1 |
NR_046007.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700031405.mm.1 |
TC0700003722.mm.1 |
MTA TC |