This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01315675_g1
See other DMTN GE Assays ›
Gene Symbol:
Dmtn
Gene Name:
dematin actin binding protein
Gene Aliases:
AI325486, Epb4.9, Epb49, dematin
Chromosome Location:
Chr.14: 70602184 - 70636048 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01315675_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dmtn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252662.1 NP_001239591.1 6 - 7 873 63
NM_001252663.1 NP_001239592.1 6 - 7 873 63
NM_001252664.1 NP_001239593.1 5 - 6 700 63
NM_001252665.1 NP_001239594.1 5 - 6 599 63
NM_001252666.1 NP_001239595.1 4 - 5 632 63
NM_013514.4 NP_038542.1 7 - 8 814 63
GenBank mRNA AF079846.3 - 5 - 6 543 63
AF155547.1 - 5 - 6 543 63
AK149301.1 - 5 - 6 510 63
AK155927.1 - 6 - 7 868 63
AK158744.1 - 5 - 6 631 63
AK162866.1 - 5 - 6 551 63
AK165204.1 - 866 63
AK165294.1 - 6 - 7 865 63
BC016897.1 - 7 - 8 814 4240747 63
BC037021.1 - 4 - 5 619 5366040 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002346.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dmtn

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
spectrin binding
protein self-association

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252662.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252662.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252663.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252663.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252664.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252664.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252665.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252665.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252666.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_001252666.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_013514.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 NM_013514.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400018357.mm.1 TC1400002346.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0