This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01316781_g1
See other EPHX1 GE Assays ›
Gene Symbol:
Ephx1
Gene Name:
epoxide hydrolase 1, microsomal
Gene Aliases:
AI195553, Eph-1, Eph1, Epxh1, mEH
Chromosome Location:
Chr.1: 180989556 - 181017495 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01316781_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ephx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010145.2 NP_034275.1 5 - 6 795 135
GenBank mRNA AK018249.1 - 5 - 6 804 135
BC029636.1 - 5 - 6 794 3491966 135
BC045194.1 - 5 - 6 795 4986422 135
BC061493.1 - 5 - 6 821 4488885 135
U89491.1 - 5 - 6 764 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003679.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ephx1

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

cellular aromatic compound metabolic process
response to toxic substance
response to organic cyclic compound
aromatic compound catabolic process
catalytic activity
epoxide hydrolase activity
hydrolase activity
enzyme binding
cis-stilbene-oxide hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100030232.mm.1 NM_010145.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100030232.mm.1 TC0100003679.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100030231.mm.1 NM_010145.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100030231.mm.1 TC0100003679.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline