This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01316809_m1
See other PRKACB GE Assays ›
Gene Symbol:
Prkacb
Gene Name:
protein kinase, cAMP dependent, catalytic, beta
Gene Aliases:
Pkacb
Chromosome Location:
Chr.3: 146729579 - 146812946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01316809_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkacb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164198.1 NP_001157670.1 1 - 2 309 109
GenBank mRNA CB723114.1 - 1 - 2 309 109

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001164198.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
protein phosphorylation
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
phosphorylation
negative regulation of meiotic cell cycle
regulation of protein processing
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
ubiquitin protein ligase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300024454.mm.1 NM_001164198.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024454.mm.1 NM_001164198.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024454.mm.1 TC0300003106.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline