This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01316923_g1
See other NDST3 GE Assays ›
Gene Symbol:
Ndst3
Gene Name:
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3
Gene Aliases:
4921531K01Rik, 4930511P15Rik, N-HSST 3, NDST-3
Chromosome Location:
Chr.3: 123526166 - 123690852 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01316923_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndst3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293682.1 NP_001280611.1 1 - 2 147 89
GenBank mRNA AK015768.1 - 1 - 2 147 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK015768.1, NM_001293682.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Protease Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
catalytic activity
GPI-anchor transamidase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
[heparan sulfate]-glucosamine N-sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
hydrolase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
deacetylase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity
hydroxyjasmonate sulfotransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline