This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01316935_m1
See other EXTL2 GE Assays ›
Gene Symbol:
Extl2
Gene Name:
exostoses (multiple)-like 2
Gene Aliases:
3000001D04Rik, AW146439
Chromosome Location:
Chr.3: 116007446 - 116029017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01316935_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Extl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021388.4 NP_067363.3 1 - 2 441 126
GenBank mRNA AB032170.1 - 1 - 2 251 126
AK028677.1 - 1 - 2 414 126
AK159281.1 - 1 - 2 428 126
BC031438.1 - 1 - 2 405 4024555 126
BC094444.1 - 1 - 2 154 4507232 126
BG694725.1 - 1 - 2 441 126

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB032170.1, AK028677.1, AK159281.1, BC031438.1, BC094444.1, NM_021388.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Extl2

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
N-acetylglucosamine metabolic process
heparan sulfate proteoglycan biosynthetic process
UDP-N-acetylgalactosamine metabolic process
glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
alpha-1,4-N-acetylgalactosaminyltransferase activity
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300009693.mm.1 NM_001163515.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009693.mm.1 NM_001163515.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009693.mm.1 NM_021388.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009693.mm.1 NM_021388.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300009693.mm.1 TC0300001185.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300009694.mm.1 NM_001163515.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009694.mm.1 NM_001163515.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009694.mm.1 NM_021388.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009694.mm.1 NM_021388.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300009694.mm.1 TC0300001185.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300009692.mm.1 NM_001163515.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009692.mm.1 NM_001163515.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009692.mm.1 NM_021388.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009692.mm.1 NM_021388.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300009692.mm.1 TC0300001185.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300009695.mm.1 NM_001163515.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009695.mm.1 NM_001163515.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009695.mm.1 NM_021388.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009695.mm.1 NM_021388.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300009695.mm.1 TC0300001185.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300009700.mm.1 NM_001163514.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009700.mm.1 NM_001163514.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009700.mm.1 NM_021388.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300009700.mm.1 NM_021388.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300009700.mm.1 TC0300001185.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 460831 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0