This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317041_m1
See other EXTL3 GE Assays ›
Gene Symbol:
Extl3
Gene Name:
exostoses (multiple)-like 3
Gene Aliases:
2900009G18Rik, AU040105, Ext1l, mKIAA0519
Chromosome Location:
Chr.14: 65052059 - 65149899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317041_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Extl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018788.3 NP_061258.2 5 - 6 3088 90
GenBank mRNA AF083550.1 - 3 - 4 2878 90
AK137842.1 - 3 - 4 2165 90
AK220344.1 - 3 - 4 2431 90
BC058526.1 - 3 - 4 2479 90
BC065073.1 - 3 - 4 2553 6834580 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002248.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Extl3

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
positive regulation of cell growth
glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
receptor activity
transferase activity
transferase activity, transferring glycosyl groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017356.mm.1 NM_018788.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017356.mm.1 NM_018788.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017356.mm.1 TC1400002248.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0