This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317292_g1
See other NEXN GE Assays ›
Gene Symbol:
Nexn
Gene Name:
nexilin
Gene Aliases:
1110046H09Rik, AA553326, NELIN
Chromosome Location:
Chr.3: 152234785 - 152266349 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317292_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nexn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199465.2 NP_955759.2 3 - 4 264 113
GenBank mRNA AK136097.1 - 3 - 4 390 113
AK136102.1 - 3 - 4 192 113
AK163414.1 - 3 - 4 264 113
AK163848.1 - 2 - 3 178 113
BC052878.1 - 3 - 4 211 30046316 113
HQ179982.1 - 3 - 4 298 113
HQ179983.1 - 3 - 4 298 113
HQ179984.1 - 3 - 4 298 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003140.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nexn

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of cell migration
cardiac muscle fiber development
regulation of cytoskeleton organization
actin binding
calmodulin-dependent protein kinase activity
structural constituent of muscle
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300024659.mm.1 NM_199465.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024659.mm.1 NM_199465.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300024659.mm.1 TC0300003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300024658.mm.1 NM_199465.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024658.mm.1 NM_199465.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300024658.mm.1 TC0300003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300024655.mm.1 NM_199465.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024655.mm.1 NM_199465.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300024655.mm.1 TC0300003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300024656.mm.1 NM_199465.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024656.mm.1 NM_199465.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300024656.mm.1 TC0300003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300024660.mm.1 NM_199465.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024660.mm.1 NM_199465.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300024660.mm.1 TC0300003140.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0