This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317296_m1
See other NEXN GE Assays ›
Gene Symbol:
Nexn
Gene Name:
nexilin
Gene Aliases:
1110046H09Rik, AA553326, NELIN
Chromosome Location:
Chr.3: 152234785 - 152266349 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317296_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nexn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199465.2 NP_955759.2 7 - 8 815 110
GenBank mRNA AK136102.1 - 7 - 8 743 110
AK162232.1 - 2 - 3 256 109
AK163414.1 - 7 - 8 815 110
AK163848.1 - 6 - 7 729 111
BC052878.1 - 7 - 8 762 30046316 110
HQ179982.1 - 7 - 8 849 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003140.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nexn

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of cell migration
cardiac muscle fiber development
regulation of cytoskeleton organization
actin binding
calmodulin-dependent protein kinase activity
structural constituent of muscle
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300024650.mm.1 NM_199465.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024650.mm.1 NM_199465.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300024650.mm.1 TC0300003140.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300024651.mm.1 NM_199465.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300024651.mm.1 NM_199465.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300024651.mm.1 TC0300003140.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0