This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317730_m1
See other BANK1 GE Assays ›
Gene Symbol:
Bank1
Gene Name:
B cell scaffold protein with ankyrin repeats 1
Gene Aliases:
A530094C12Rik, AI451642, AVIEF, BANK
Chromosome Location:
Chr.3: 136053363 - 136326141 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317730_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bank1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033350.2 NP_001028522.2 13 - 14 2309 68
GenBank mRNA AK041039.1 - 4 - 5 327 68
AK148665.1 - 2 - 3 104 68
AK156201.1 - 13 - 14 2319 68
AY178734.1 - 13 - 14 2329 68
BC116328.1 - 3 - 4 392 68
BE848854.1 - 4 - 5 463 68
EU051378.1 - 12 - 13 1910 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bank1

Gene Ontology Categories:

Function(s) Process(es)

B cell activation
positive regulation of MAPK cascade
negative regulation of translational initiation
negative regulation of B cell activation
negative regulation of protein kinase B signaling
negative regulation of interleukin-6 secretion
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0