This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317733_m1
See other BANK1 GE Assays ›
Gene Symbol:
Bank1
Gene Name:
B cell scaffold protein with ankyrin repeats 1
Gene Aliases:
A530094C12Rik, AI451642, AVIEF, BANK
Chromosome Location:
Chr.3: 136053363 - 136326141 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317733_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bank1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033350.2 NP_001028522.2 1 - 2 179 77
GenBank mRNA AK041242.1 - 1 - 2 140 77
AK156201.1 - 1 - 2 189 77
AY178734.1 - 1 - 2 199 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002998.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bank1

Gene Ontology Categories:

Function(s) Process(es)

B cell activation
positive regulation of MAPK cascade
negative regulation of translational initiation
negative regulation of B cell activation
negative regulation of protein kinase B signaling
negative regulation of interleukin-6 secretion
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023691.mm.1 NM_001033350.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300023691.mm.1 TC0300002998.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023689.mm.1 NM_001033350.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300023689.mm.1 TC0300002998.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023690.mm.1 NM_001033350.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300023690.mm.1 TC0300002998.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0