This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317783_m1
See other SLCO3A1 GE Assays ›
Gene Symbol:
Slco3a1
Gene Name:
solute carrier organic anion transporter family, member 3a1
Gene Aliases:
5830414C08Rik, Anr1, MJAM, OATP-D, R75096, Slc21a11
Chromosome Location:
Chr.7: 74275418 - 74554780 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317783_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco3a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038643.1 NP_001033732.1 2 - 3 829 120
NM_023908.2 NP_076397.2 2 - 3 829 120
GenBank mRNA AF226324.1 - 2 - 3 727 120
AK037938.1 - 2 - 3 644 120
AK151781.1 - 2 - 3 799 120
AK152488.1 - 2 - 3 663 120
AK166266.1 - 2 - 3 120
BC025059.1 - 2 - 3 662 2598913 120
BC094464.1 - 2 - 3 621 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003517.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco3a1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
oligopeptide transport
organic anion transport
prostaglandin transport
anion transmembrane transport
transporter activity
organic anion transmembrane transporter activity
oligopeptide transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700029801.mm.1 NM_001038643.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029801.mm.1 NM_001038643.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029801.mm.1 NM_023908.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029801.mm.1 NM_023908.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700029801.mm.1 TC0700003517.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029802.mm.1 NM_001038643.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029802.mm.1 NM_001038643.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029802.mm.1 NM_023908.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029802.mm.1 NM_023908.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700029802.mm.1 TC0700003517.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029803.mm.1 NM_001038643.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029803.mm.1 NM_001038643.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029803.mm.1 NM_023908.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029803.mm.1 NM_023908.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700029803.mm.1 TC0700003517.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029804.mm.1 NM_001038643.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029804.mm.1 NM_001038643.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029804.mm.1 NM_023908.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029804.mm.1 NM_023908.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700029804.mm.1 TC0700003517.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029798.mm.1 NM_001038643.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029798.mm.1 NM_001038643.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029798.mm.1 NM_023908.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029798.mm.1 NM_023908.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700029798.mm.1 TC0700003517.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0