This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317943_g1
See other STARD5 GE Assays ›
Gene Symbol:
Stard5
Gene Name:
StAR-related lipid transfer (START) domain containing 5
Gene Aliases:
2310058G22Rik, AI607880, D7Ertd152e
Chromosome Location:
Chr.7: 83632017 - 83642328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317943_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023377.4 NP_075866.2 1 - 2 122 59
GenBank mRNA AF480302.1 - 1 - 2 98 59
AK009980.1 - 1 - 2 128 59
AK077328.1 - 1 - 2 140 59
AK080060.1 - 1 - 2 140 59
AK085852.1 - 1 - 2 135 59
AK142503.1 - 1 - 2 140 59
AK158178.1 - 1 - 2 124 59
AY007808.1 - 1 - 2 117 59
BC031733.1 - 1 - 2 126 59
BC058226.1 - 1 - 2 122 6308417 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK077328.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard5

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
lipid binding
cholesterol binding
bile acid binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700010611.mm.1 TC0700001198.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700010611.mm.1 NM_023377.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700010611.mm.1 NM_023377.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700010609.mm.1 TC0700001198.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700010609.mm.1 NM_023377.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700010609.mm.1 NM_023377.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 390680 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0