This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01317944_g1
See other STARD5 GE Assays ›
Gene Symbol:
Stard5
Gene Name:
StAR-related lipid transfer (START) domain containing 5
Gene Aliases:
2310058G22Rik, AI607880, D7Ertd152e
Chromosome Location:
Chr.7: 83632017 - 83642328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01317944_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023377.4 NP_075866.2 2 - 3 172 81
GenBank mRNA AF480302.1 - 2 - 3 148 81
AK009980.1 - 2 - 3 178 81
AK077328.1 - 2 - 3 190 81
AK080060.1 - 2 - 3 190 81
AK085852.1 - 2 - 3 185 81
AK142503.1 - 2 - 3 190 81
AK158178.1 - 2 - 3 174 81
AY007808.1 - 2 - 3 167 81
BC031733.1 - 2 - 3 176 81
BC058226.1 - 2 - 3 172 6308417 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK077328.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard5

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
lipid binding
cholesterol binding
bile acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700010612.mm.1 TC0700001198.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700010612.mm.1 NM_023377.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700010612.mm.1 NM_023377.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0