This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318157_m1
See other GSTCD GE Assays ›
Gene Symbol:
Gstcd
Gene Name:
glutathione S-transferase, C-terminal domain containing
Gene Aliases:
4933434L15Rik
Chromosome Location:
Chr.3: 132981752 - 133091971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318157_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gstcd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026231.2 NP_080507.2 2 - 3 608 93
GenBank mRNA AK020021.1 - 1 - 2 218 93
AK029724.1 - 2 - 3 607 93
AK033409.1 - 2 - 3 646 93
AK037169.1 - 2 - 3 647 93
AK044441.1 - 2 - 3 616 93
BC047136.1 - 2 - 3 608 6311866 93
BC061240.1 - 2 - 3 604 6743615 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002973.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gstcd

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023478.mm.1 NM_026231.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023478.mm.1 NM_026231.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300023478.mm.1 TC0300002973.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300023481.mm.1 NM_026231.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023481.mm.1 NM_026231.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300023481.mm.1 TC0300002973.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 929082 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0