This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318160_m1
See other GSTCD GE Assays ›
Gene Symbol:
Gstcd
Gene Name:
glutathione S-transferase, C-terminal domain containing
Gene Aliases:
4933434L15Rik
Chromosome Location:
Chr.3: 132981752 - 133091971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318160_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gstcd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026231.2 NP_080507.2 5 - 6 1424 85
GenBank mRNA AK017059.1 - 3 - 4 699 85
AK020021.1 - 4 - 5 1034 85
AK029724.1 - 5 - 6 1423 85
AK033409.1 - 5 - 6 1462 85
AK044441.1 - 5 - 6 1432 85
BC047136.1 - 5 - 6 1424 6311866 85
BC061240.1 - 5 - 6 1420 6743615 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002973.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gstcd

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300023472.mm.1 NM_026231.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300023472.mm.1 NM_026231.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300023472.mm.1 TC0300002973.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline