This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318201_m1
See other ETNPPL GE Assays ›
Gene Symbol:
Etnppl
Gene Name:
ethanolamine phosphate phospholyase
Gene Aliases:
1300019H02Rik, AI195447, Agxt2l1
Chromosome Location:
Chr.3: 130617448 - 130635750 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318201_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etnppl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163587.1 NP_001157059.1 12 - 13 1379 80
NM_027907.3 NP_082183.2 11 - 12 1417 80
GenBank mRNA AK005060.1 - 11 - 12 1418 80
AK049937.1 - 12 - 13 1379 80
BC043680.1 - 11 - 12 1404 5101568 80
BC058592.1 - 11 - 12 1379 5363337 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001322.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etnppl

Gene Ontology Categories:

Function(s) Process(es)

biological_process
catalytic activity
transaminase activity
transferase activity
lyase activity
pyridoxal phosphate binding
ethanolamine-phosphate phospho-lyase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300010723.mm.1 NM_001163587.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300010723.mm.1 NM_001163587.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300010723.mm.1 NM_027907.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300010723.mm.1 NM_027907.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300010723.mm.1 TC0300001322.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0