This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318462_m1
See other IGF1R GE Assays ›
Gene Symbol:
Igf1r
Gene Name:
insulin-like growth factor I receptor
Gene Aliases:
A330103N21Rik, CD221, D930020L01, IGF-1R, hyft
Chromosome Location:
Chr.7: 67952257 - 68233668 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318462_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igf1r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010513.2 NP_034643.2 4 - 5 1708 78
GenBank mRNA AF056187.1 - 4 - 5 1106 78
AK155079.1 - 3 - 4 586 78
AK155884.1 - 3 - 4 604 78
BC138868.1 - 4 - 5 1138 8861890 78
BC138869.1 - 4 - 5 1138 8861891 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igf1r

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
immune response
transmembrane receptor protein tyrosine kinase signaling pathway
axonogenesis
brain development
epidermis development
organ morphogenesis
negative regulation of muscle cell apoptotic process
phosphorylation
establishment of cell polarity
male sex determination
positive regulation of cell migration
mammary gland development
exocrine pancreas development
neuron projection development
positive regulation of cytokinesis
peptidyl-tyrosine autophosphorylation
negative regulation of apoptotic process
negative regulation of MAPK cascade
positive regulation of MAPK cascade
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of DNA replication
positive regulation of mitotic nuclear division
regulation of JNK cascade
protein autophosphorylation
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of DNA metabolic process
protein tetramerization
protein heterooligomerization
inactivation of MAPKK activity
positive regulation of protein kinase B signaling
negative regulation of protein kinase B signaling
prostate gland epithelium morphogenesis
positive regulation of steroid hormone biosynthetic process
nucleotide binding
G-protein alpha-subunit binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
insulin-like growth factor-activated receptor activity
insulin receptor binding
protein binding
insulin-like growth factor binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
insulin-like growth factor I binding
identical protein binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline