This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318601_m1
See other SV2B GE Assays ›
Gene Symbol:
Sv2b
Gene Name:
synaptic vesicle glycoprotein 2 b
Gene Aliases:
A830038F04Rik, AI850768, mKIAA0735
Chromosome Location:
Chr.7: 75114890 - 75309262 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318601_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sv2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109753.1 NP_001103223.1 1 - 2 180 97
GenBank mRNA AK035927.1 - 1 - 2 105 97
AK043830.1 - 1 - 2 107 97
AK139478.1 - 1 - 2 82 97
AK140104.1 - 1 - 2 100 97
AK147261.1 - 1 - 2 174 97
BY269935.1 - 1 - 2 181 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003526.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK043830.1,AK139478.1,AK140104.1,AK147261.1,NM_001109753.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
neurotransmitter transport
chemical synaptic transmission
transmembrane transport
transporter activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700029849.mm.1 NM_001109753.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029849.mm.1 NM_001109753.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029849.mm.1 NM_153579.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029849.mm.1 NM_153579.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700029849.mm.1 TC0700003526.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029850.mm.1 NM_001109753.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029850.mm.1 NM_001109753.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029850.mm.1 NM_153579.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029850.mm.1 NM_153579.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700029850.mm.1 TC0700003526.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029851.mm.1 NM_001109753.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029851.mm.1 NM_001109753.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029851.mm.1 TC0700003526.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 543542 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0