This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318604_m1
See other SV2B GE Assays ›
Gene Symbol:
Sv2b
Gene Name:
synaptic vesicle glycoprotein 2 b
Gene Aliases:
A830038F04Rik, AI850768, mKIAA0735
Chromosome Location:
Chr.7: 75114890 - 75309262 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318604_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sv2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109753.1 NP_001103223.1 4 - 5 1350 62
NM_153579.4 NP_705807.2 4 - 5 1557 62
GenBank mRNA AK029736.1 - 5 - 6 1382 62
AK043830.1 - 4 - 5 1277 62
AK122359.1 - 3 - 4 502 62
AK139478.1 - 4 - 5 1252 62
AK140104.1 - 4 - 5 1270 62
AK147261.1 - 4 - 5 1344 62
AK159028.1 - 4 - 5 1255 62
BC060224.1 - 3 - 4 941 30356990 62
BC079568.1 - 4 - 5 1557 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003526.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sv2b

Gene Ontology Categories:

Function(s) Process(es)

transport
neurotransmitter transport
chemical synaptic transmission
transmembrane transport
transporter activity
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700029844.mm.1 NM_001109753.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029844.mm.1 NM_001109753.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029844.mm.1 NM_153579.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029844.mm.1 NM_153579.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700029844.mm.1 TC0700003526.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700029845.mm.1 NM_001109753.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029845.mm.1 NM_001109753.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029845.mm.1 NM_153579.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700029845.mm.1 NM_153579.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700029845.mm.1 TC0700003526.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0