This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318861_m1
See other UBE2D3 GE Assays ›
Gene Symbol:
Ube2d3
Gene Name:
ubiquitin-conjugating enzyme E2D 3
Gene Aliases:
1100001F19Rik, 9430029A22Rik, AA414951
Chromosome Location:
Chr.3: 135438244 - 135467180 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318861_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2d3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK077666.1 - 1 - 2 168 110
AK151295.1 - 1 - 2 222 110
AK153100.1 - 1 - 2 222 110
AK153423.1 - 1 - 2 168 110
AK159125.1 - 1 - 2 168 110

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK077666.1, AK151295.1, AK153100.1, AK159125.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline