This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01318975_g1
See other SLC7A8 GE Assays ›
Gene Symbol:
Slc7a8
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 8
Gene Aliases:
AA408822, LAT2
Chromosome Location:
Chr.14: 54722215 - 54781886 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01318975_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016972.2 NP_058668.1 5 - 6 1339 86
GenBank mRNA AF171668.1 - 5 - 6 994 86
BC059004.1 - 5 - 6 1339 6849059 86
Y19022.1 - 5 - 6 1006 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002067.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a8

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
organic cation transport
neutral amino acid transport
L-amino acid transport
amine transport
transmembrane transport
cation transmembrane transport
toxin transport
L-alpha-amino acid transmembrane transport
amine transmembrane transporter activity
organic cation transmembrane transporter activity
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity
toxin transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015610.mm.1 NM_016972.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015610.mm.1 NM_016972.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400015610.mm.1 TC1400002067.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0