This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319219_m1
See other PRMT5 GE Assays ›
Gene Symbol:
Prmt5
Gene Name:
protein arginine N-methyltransferase 5
Gene Aliases:
Jbp1, Skb1
Chromosome Location:
Chr.14: 54507182 - 54517470 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319219_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prmt5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013768.3 NP_038796.2 7 - 8 895 117
GenBank mRNA AF167573.1 - 7 - 8 763 117
AK076346.1 - 7 - 8 786 117
AK132414.1 - 7 - 8 787 117
AK165165.1 - 7 - 8 813 117
BC023905.1 - 7 - 8 842 5321370 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002057.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prmt5

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
spliceosomal snRNP assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
protein methylation
chromatin modification
peptidyl-arginine methylation
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
methylation
circadian regulation of gene expression
histone arginine methylation
peptidyl-arginine N-methylation
endothelial cell activation
histone H4-R3 methylation
regulation of DNA methylation
negative regulation of cell differentiation
rhythmic process
positive regulation of oligodendrocyte differentiation
regulation of ERK1 and ERK2 cascade
rRNA (adenine-N6,N6-)-dimethyltransferase activity
core promoter sequence-specific DNA binding
chromatin binding
transcription corepressor activity
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
protein binding
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
histone-arginine N-methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
protein complex binding
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
protein-arginine omega-N symmetric methyltransferase activity
ribonucleoprotein complex binding
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
protein heterodimerization activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015448.mm.1 NM_013768.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015448.mm.1 NM_013768.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015448.mm.1 TC1400002057.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015449.mm.1 NM_013768.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015449.mm.1 NM_013768.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015449.mm.1 TC1400002057.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015447.mm.1 NM_013768.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015447.mm.1 NM_013768.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015447.mm.1 TC1400002057.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015450.mm.1 NM_013768.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015450.mm.1 NM_013768.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015450.mm.1 TC1400002057.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015451.mm.1 NM_013768.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015451.mm.1 NM_013768.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015451.mm.1 TC1400002057.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0